Learn More
ACROBiosystems FITC-Labeled Human CEACAM-5 / CD66e Protein, His Tag (MALS verified)
Catalog No. 502054997
Encompass_Preferred
Recombinant Protein;200UG;FITC-Labeled Human CEACAM-5, His Tag (CE5-HF254) is expressed from human 293 cells (HEK293). It contains AA Lys 35 - Ala 685 (Accession # P06731-1). It is the FITC labeled form of Human CEACAM-5, His Tag (Cat. No.CE5-H5226).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 72.2kDa. The protein migrates as 90-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-CEACAM-5 (labetuzumab) Antibody, Human IgG1 at 1 μg/mL (100 μL/well) can bind FITC-Labeled Human CEACAM-5, His Tag (Cat. No. CE5-HF254) with a linear range of 0.039-0.625 μg/mL (QC tested).
Catalog No. 50-205-4997 Supplier ACROBiosystems Supplier No. CE5HF254200UG
Recombinant Protein;200UG;FITC-Labeled Human CEACAM-5, His Tag (CE5-HF254) is expressed from human 293 cells (HEK293). It contains AA Lys 35 - Ala 685 (Accession # P06731-1). It is the FITC labeled form of Human CEACAM-5, His Tag (Cat. No.CE5-H5226).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 72.2kDa. The protein migrates as 90-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-CEACAM-5 (labetuzumab) Antibody, Human IgG1 at 1 μg/mL (100 μL/well) can bind FITC-Labeled Human CEACAM-5, His Tag (Cat. No. CE5-HF254) with a linear range of 0.039-0.625 μg/mL (QC tested).