Learn More
ACROBiosystems ACROBiosystems FITC-Labeled Human DLL3 Protein, Fc Tag
Supplier:  ACROBiosystems DL3HF25325UG
Encompass_Preferred
Recombinant Protein;25UG;FITC-Labeled Human DLL3, Fc Tag (DL3-HF253) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Leu 492 (Accession # Q9NYJ7-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 75.2 kDa. The protein migrates as 60-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-DLL3 Antibody, Human IgG1 at 2 μg/mL (100 μL/well) can bind FITC-Labeled Human DLL3, Fc Tag (Cat. No. DL3-HF253) with a linear range of 0.01-0.313 μg/mL (QC tested).
Catalog No. 50-210-6365