Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems FITC-Labeled Human DLL3 Protein, His Tag, 25 μg

Catalog No. 502106362
Encompass_Preferred

FITC-labeled human DLL3 protein is a transmembrane protein from the Delta/Serrate/Lag-2 (DSL) family of Notch ligands. It plays a role in the formation of somite boundaries during segmentation of the paraxial mesoderm and may be required to divert neurons along a specific differentiation pathway. This protein is expressed on the surface of tumor cells but not in normal adult tissues.

  • Expressed from human 293 cells with a C-terminal polyhistidine tag
  • FITC conjugated using standard chemical labeling with a molar ratio of 1.5 to 3.5
  • Purity ≥90% as determined by SDS-PAGE
  • Lyophilized from 0.22 μm filtered PBS and arginine solution, pH 7.4, with trehalose
  • Excitation wavelength of 488 nm and emission wavelength of 535 nm

Catalog No. 50-210-6362 Supplier ACROBiosystems Supplier No. DL3HF2H425UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FITC-labeled human DLL3 protein is a transmembrane protein from the Delta/Serrate/Lag-2 (DSL) family of Notch ligands. It plays a role in the formation of somite boundaries during segmentation of the paraxial mesoderm and may be required to divert neurons along a specific differentiation pathway. This protein is expressed on the surface of tumor cells but not in normal adult tissues.

  • Expressed from human 293 cells with a C-terminal polyhistidine tag
  • FITC conjugated using standard chemical labeling with a molar ratio of 1.5 to 3.5
  • Purity ≥90% as determined by SDS-PAGE
  • Lyophilized from 0.22 μm filtered PBS and arginine solution, pH 7.4, with trehalose
  • Excitation wavelength of 488 nm and emission wavelength of 535 nm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.