Learn More
ACROBiosystems FITC-Labeled Human EMMPRIN / CD147 Protein, His Tag
Catalog No. 502054959
Encompass_Preferred
Recombinant Protein;25UG;FITC-Labeled Human EMMPRIN, His Tag (CD7-HF222) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - His 205 (Accession # P35613-2). It is the FITC labeled form of Human EMMPRIN, His Tag (CD7-H5222).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 20.9 kDa. The protein migrates as 28-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-205-4959 Supplier ACROBiosystems Supplier No. CD7HF22225UG
Recombinant Protein;25UG;FITC-Labeled Human EMMPRIN, His Tag (CD7-HF222) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - His 205 (Accession # P35613-2). It is the FITC labeled form of Human EMMPRIN, His Tag (CD7-H5222).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 20.9 kDa. The protein migrates as 28-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.