Learn More
ACROBiosystems FITC-Labeled Human EpCAM / TROP1 Protein, His Tag
Catalog No. 502055124
Encompass_Preferred
Recombinant Protein;200UG;FITC-Labeled Human EpCAM, His Tag (EPM-HF225) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession # AAH14785.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.2 kDa.
Catalog No. 50-205-5124 Supplier ACROBiosystems Supplier No. EPMHF225200UG
Recombinant Protein;200UG;FITC-Labeled Human EpCAM, His Tag (EPM-HF225) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession # AAH14785.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.2 kDa.