Learn More
ACROBiosystems FITC-Labeled Human EpCAM / TROP1 Protein, Fc Tag
Catalog No. 502055127
Encompass_Preferred
Recombinant Protein;25UG;FITC-Labeled Human EpCAM, Fc Tag (EPM-HF255) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession # AAH14785.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 53.5 kDa.
Catalog No. 50-205-5127 Supplier ACROBiosystems Supplier No. EPMHF25525UG
Recombinant Protein;25UG;FITC-Labeled Human EpCAM, Fc Tag (EPM-HF255) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession # AAH14785.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 53.5 kDa.