Learn More
ACROBiosystems ACROBiosystems FITC-Labeled Human LILRB4 / CD85k / ILT3 Protein, His Tag
Catalog No. 502106290
Encompass_Preferred
Recombinant Protein;200UG;FITC-Labeled Human LILRB4, His Tag (LI4-HF2H3) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Glu 259 (Accession # AAH26309.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.2 kDa. The protein migrates as 30-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6290 Supplier ACROBiosystems Supplier No. LI4HF2H3200UG
Recombinant Protein;200UG;FITC-Labeled Human LILRB4, His Tag (LI4-HF2H3) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Glu 259 (Accession # AAH26309.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.2 kDa. The protein migrates as 30-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.