Learn More
ACROBiosystems ACROBiosystems FITC-Labeled Human NCAM-1 / CD56 Protein, His Tag
Supplier:  ACROBiosystems NC1HF2H525UG
Encompass_Preferred
Recombinant Protein;25UG;FITC-Labeled Human NCAM-1, His Tag (NC1-HF2H5) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Gly 718 (Accession # P13591-2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 79.3 kDa. The protein migrates as 90-116 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6192