Learn More
ACROBiosystems ACROBiosystems FITC-Labeled Human NCAM-1 / CD56 Protein, Fc Tag
Catalog No. 502106194
Encompass_Preferred
Recombinant Protein;25UG;FITC-Labeled Human NCAM-1, Fc Tag (NC1-HF256) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Gly 718 (Accession # P13591-2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 104.0 kDa. The protein migrates as 125-150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-NCAM-1 Antibody, Human IgG1 at 5 μg/mL (100 μL/well) can bind FITC-Labeled Human NCAM-1, Fc Tag (Cat. No. NC1-HF256) with a linear range of 0.08-0.625 μg/mL (QC tested).
Catalog No. 50-210-6194 Supplier ACROBiosystems Supplier No. NC1HF25625UG
Recombinant Protein;25UG;FITC-Labeled Human NCAM-1, Fc Tag (NC1-HF256) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Gly 718 (Accession # P13591-2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 104.0 kDa. The protein migrates as 125-150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-NCAM-1 Antibody, Human IgG1 at 5 μg/mL (100 μL/well) can bind FITC-Labeled Human NCAM-1, Fc Tag (Cat. No. NC1-HF256) with a linear range of 0.08-0.625 μg/mL (QC tested).