Learn More
ACROBiosystems FITC-Labeled Human Nectin-4 Protein, His Tag
Catalog No. 502054957
Encompass_Preferred
Recombinant Protein;25UG;FITC-Labeled Human Nectin-4, His Tag (NE4-HF2H3) is expressed from human 293 cells (HEK293). It contains AA Gly 32 - Ser 349 (Accession # Q96NY8-1). It is the FITC labeled form of Human Nectin-4, His Tag (NE4-H52H3).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.0 kDa.
Catalog No. 50-205-4957 Supplier ACROBiosystems Supplier No. NE4HF2H325UG
Recombinant Protein;25UG;FITC-Labeled Human Nectin-4, His Tag (NE4-HF2H3) is expressed from human 293 cells (HEK293). It contains AA Gly 32 - Ser 349 (Accession # Q96NY8-1). It is the FITC labeled form of Human Nectin-4, His Tag (NE4-H52H3).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.0 kDa.