Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems FITC-Labeled Human Respiratory Syncytial Virus A (Strain A2) Pre-F0 Protein, His Tag, 50 μg

Catalog No. 502370195
Encompass_Preferred

FITC-labeled human respiratory syncytial virus A (strain A2) Pre-F0 protein is a fluorescent recombinant pre-fusion F0 glycoprotein produced in HEK293 cells, supplied lyophilized as a 50 μg vial and intended for use in fluorescence-based binding and detection assays.

  • Size: 50 μg.
  • Tag: C-terminal polyhistidine (His) tag.
  • Expression system: Human 293 cells (HEK293).
  • Amino acid range: Gln 26-Leu 513.
  • Fluorescent label: FITC (excitation 488 nm, emission 535 nm).
  • Purity: >90% by SDS-PAGE.
  • FITC to protein molar ratio: 1.0-2.0.
  • Molecular weight: Calculated 56.1 kDa; migrates 60-65 kDa under reducing SDS-PAGE due to glycosylation.
  • Formulation and storage: Lyophilized from PBS, pH 7.4, with trehalose; store lyophilized at -20°C or lower; protect from light and avoid repeated freeze-thaw cycles.

Catalog No. 50-237-0195 Supplier ACROBiosystems Supplier No. RSFV52H350UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FITC-labeled human respiratory syncytial virus A (strain A2) Pre-F0 protein is a fluorescent recombinant pre-fusion F0 glycoprotein produced in HEK293 cells, supplied lyophilized as a 50 μg vial and intended for use in fluorescence-based binding and detection assays.

  • Size: 50 μg.
  • Tag: C-terminal polyhistidine (His) tag.
  • Expression system: Human 293 cells (HEK293).
  • Amino acid range: Gln 26-Leu 513.
  • Fluorescent label: FITC (excitation 488 nm, emission 535 nm).
  • Purity: >90% by SDS-PAGE.
  • FITC to protein molar ratio: 1.0-2.0.
  • Molecular weight: Calculated 56.1 kDa; migrates 60-65 kDa under reducing SDS-PAGE due to glycosylation.
  • Formulation and storage: Lyophilized from PBS, pH 7.4, with trehalose; store lyophilized at -20°C or lower; protect from light and avoid repeated freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.