Learn More
ACROBiosystems ACROBiosystems FITC-Labeled Human uPAR / PLAUR Protein, His Tag
Catalog No. 502106292
Encompass_Preferred
Recombinant Protein;200UG;FITC-Labeled Human uPAR, His Tag (UPR-HF2H3) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Arg 303 (Accession # Q03405-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 32.3 kDa. The protein migrates as 45-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-uPAR Antibody, Mouse IgG1 at 1 μg/mL (100 μL/well) can bind FITC-Labeled Human uPAR, His Tag (Cat. No. UPR-HF2H3) with a linear range of 39-156 ng/mL (QC tested).
Catalog No. 50-210-6292 Supplier ACROBiosystems Supplier No. UPRHF2H3200UG
Recombinant Protein;200UG;FITC-Labeled Human uPAR, His Tag (UPR-HF2H3) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Arg 303 (Accession # Q03405-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 32.3 kDa. The protein migrates as 45-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-uPAR Antibody, Mouse IgG1 at 1 μg/mL (100 μL/well) can bind FITC-Labeled Human uPAR, His Tag (Cat. No. UPR-HF2H3) with a linear range of 39-156 ng/mL (QC tested).