Learn More
ACROBiosystems FITC-Labeled Human VEGF R2 / KDR Protein, His Tag
Catalog No. 502054990
Encompass_Preferred
Recombinant Protein;200UG;FITC-Labeled Human VEGF R2, His Tag (VE2-HF254) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Glu 764 (Accession # AAI31823.1). It is the FITC labeled form of Human VEGF R2, His Tag (Cat. No.KDR-H5227).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 85.2 kDa. The protein migrates as 110-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-205-4990 Supplier ACROBiosystems Supplier No. VE2HF254200UG
Recombinant Protein;200UG;FITC-Labeled Human VEGF R2, His Tag (VE2-HF254) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Glu 764 (Accession # AAI31823.1). It is the FITC labeled form of Human VEGF R2, His Tag (Cat. No.KDR-H5227).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 85.2 kDa. The protein migrates as 110-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.