Learn More
ACROBiosystems ACROBiosystems FITC-Labeled Human VEGF165 Protein, His Tag
Supplier:  ACROBiosystems VE5HF24825UG
Encompass_Preferred
Recombinant Protein;25UG;FITC-Labeled Human VEGF165, His Tag (VE5-HF248) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 191 (Accession # P15692-4).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 21.1 kDa. The protein migrates as 27-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Bevacizumab at 2 μg/mL (100 μL/well) can bind FITC-Labeled Human VEGF165, His Tag (Cat. No. VE5-HF248) with a linear range of 39-156 ng/mL (QC tested).
Catalog No. 50-210-7074