Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fitc-labeled ODN 1018 sodium | 5 MG
SDP

Catalog No. 5000375713
Encompass_Preferred

FITC-labeled ODN 1018 sodium is a synthetic oligonucleotide, presented as a white to light yellow solid. This product is provided as the sodium salt, with a molecular weight of 8171.00, and is designed for research applications.

  • Appearance: White to light yellow solid
  • Purity: Up to 98.71%
  • Molecular weight: 8171.00
  • Storage: -20°C in sealed storage, away from moisture
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month

Catalog No. 50-003-75713 Supplier Medchemexpress LLC Supplier No. HY150724B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FITC-labeled ODN 1018 sodium is a synthetic oligonucleotide, presented as a white to light yellow solid. This product is provided as the sodium salt, with a molecular weight of 8171.00, and is designed for research applications.

  • Appearance: White to light yellow solid
  • Purity: Up to 98.71%
  • Molecular weight: 8171.00
  • Storage: -20°C in sealed storage, away from moisture
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.