Learn More
Medchemexpress LLC FITC-labeled ODN 1018 sodium | DNA, d(P-thio)(T-G-A-C-T-G-T-G-A-A-C-G-T-T-C-G-A-G-A-T-G-A), sodium salt | 1 MG
SDP
Catalog No. 5000375734
Encompass_Preferred
FITC-labeled ODN 1018 (sodium) is an oligodeoxynucleotide that acts as a TLR-9 agonist.
  • Evaluates CpG ODN cellular uptake.
  • Evaluates CpG ODN localization by confocal laser-scanning microscopy (excitation 495 nm, emission 520 nm) or flow cytometry.
Catalog No. 50-003-75734 Supplier Medchemexpress LLC Supplier No. HY150724B1MG
FITC-labeled ODN 1018 (sodium) is an oligodeoxynucleotide that acts as a TLR-9 agonist.
  • Evaluates CpG ODN cellular uptake.
  • Evaluates CpG ODN localization by confocal laser-scanning microscopy (excitation 495 nm, emission 520 nm) or flow cytometry.