Learn More
Medchemexpress LLC FITC-labeled ODN 2395 sodium | 1 MG
SDP
Catalog No. 5000375685
Encompass_Preferred
FITC-labeled ODN 2395 (sodium) is a C class oligodeoxynucleotide that acts as a TLR9 agonist. It is suitable for evaluating CpG ODN cellular uptake and localization.
  • Evaluate cellular uptake and localization by confocal laser-scanning microscopy (excitation 495 nm, emission 520 nm).
  • Evaluate cellular uptake and localization by flow cytometry.
  • For research use only.
Catalog No. 50-003-75685 Supplier Medchemexpress LLC Supplier No. HY150743B1MG
FITC-labeled ODN 2395 (sodium) is a C class oligodeoxynucleotide that acts as a TLR9 agonist. It is suitable for evaluating CpG ODN cellular uptake and localization.
  • Evaluate cellular uptake and localization by confocal laser-scanning microscopy (excitation 495 nm, emission 520 nm).
  • Evaluate cellular uptake and localization by flow cytometry.
  • For research use only.