Learn More
Medchemexpress LLC Fitc-labeled ODN 2395 sodium | 5 MG
SDP
Catalog No. 5000374053
Encompass_Preferred
FITC-labeled ODN 2395 (sodium) is a C class oligodeoxynucleotide that acts as a TLR9 agonist. It is intended for research use only and should not be sold to patients. This product is ideal for evaluating CpG ODN cellular uptake and localization studies.
  • Can be used to evaluate CpG ODN cellular uptake and localization
  • Suitable for confocal laser-scanning microscopy (excitation 495 nm, emission 520 nm)
  • Also suitable for flow cytometry applications
Catalog No. 50-003-74053 Supplier Medchemexpress LLC Supplier No. HY150743B5MG
FITC-labeled ODN 2395 (sodium) is a C class oligodeoxynucleotide that acts as a TLR9 agonist. It is intended for research use only and should not be sold to patients. This product is ideal for evaluating CpG ODN cellular uptake and localization studies.
  • Can be used to evaluate CpG ODN cellular uptake and localization
  • Suitable for confocal laser-scanning microscopy (excitation 495 nm, emission 520 nm)
  • Also suitable for flow cytometry applications