Learn More
Immunology Consultants FITC Conjugated Goat anti-Mouse C3 Antibody, 0.1 mg
SDP
Supplier:  Immunology Consultants GC390FZ
Encompass_Preferred
Goats were immunized with highly purified mouse C3 and the resulting antiserum was collected. Antibody was immunoaffinity purified off a mouse C3 containing immunosorbent and conjugated with fluorescein isothiocyanate (FITC). Antibody concentration was determined using an absorbance at 280 nm: 1.4equals 1.0 mg of IgG. FITC/Protein Ratio = 3.0 –6.0. This antibody will react with mouse C3 as determined by ELISA and IEP techniques. It is suitable for blotting, ELISA, and IF applications. This antibody may cross react with C3 from other species. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-224-8791