Learn More
Cytek Biosciences Inc FITC aMu KLRG1 Anti Primary
Supplier: Cytek Biosciences Inc 355893U100
Product Name: FITC Anti-Mouse KLRG1 (2F1)
Product Class: Antibody
Type: Primary
Product Name: FITC Anti-Mouse KLRG1 (2F1)
Cytek Catalog Number: 35-5893-U100
Marker: KLRG1
Target Species: Mouse
Clonality: Monoclonal
Clone: 2F1
Host Species: Golden Syrian Hamster
Isotype: IgG
Conjugation: FITC
Size: 100 μg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.