Learn More
Cytek Biosciences Inc FITC MS MHCII(M5/114.15.2)100U
Supplier:  Cytek Biosciences Inc 355321U100
Encompass_Preferred
FITC Anti-Mouse MHC Class II (I-A/I-E) (M5/114.15.2) Isotype: Rat IgG2b, kappa Apps: FC Size: 100 ug
Catalog No. 50-105-4513