Learn More
Biolegend FITC Mouse IgG1, κ isotype Ctrl
Supplier:  Biolegend 981802
Encompass_Preferred
FITC Mouse IgG1, κ isotype Ctrl [MOPC-21]; Isotype: Mouse IgG1, κ; Apps: FC; Size: 500 uL
Catalog No. 50-207-5513