Learn More
Medchemexpress LLC FITC-PEG-NHS | 98.0% | 10 MG
SDP
Catalog No. 5000297497
Encompass_Preferred
FITC-PEG-NHS (average molecular weight 3,400) is a heterobifunctional fluorescent reagent composed of fluorescein (FITC) linked to polyethylene glycol (PEG) and terminated with an N-hydroxysuccinimide (NHS) ester. The NHS ester reacts with primary amines to form stable amide bonds, enabling conjugation of the FITC fluorophore to proteins, peptides, and other amine-containing biomolecules for labeling and imaging.
  • Heterobifunctional fluorescent reagent with average molecular weight 3,400.
  • NHS ester reactive toward primary amines to form stable amide bonds.
  • Suitable for fluorescent labeling of proteins, peptides, and other amine-containing biomolecules.
  • High supplied purity (98.0%).
  • Available in lab-scale and larger package sizes to support research workflows.
Catalog No. 50-002-97497 Supplier Medchemexpress LLC Supplier No. HYD2801B10MG
FITC-PEG-NHS (average molecular weight 3,400) is a heterobifunctional fluorescent reagent composed of fluorescein (FITC) linked to polyethylene glycol (PEG) and terminated with an N-hydroxysuccinimide (NHS) ester. The NHS ester reacts with primary amines to form stable amide bonds, enabling conjugation of the FITC fluorophore to proteins, peptides, and other amine-containing biomolecules for labeling and imaging.
  • Heterobifunctional fluorescent reagent with average molecular weight 3,400.
  • NHS ester reactive toward primary amines to form stable amide bonds.
  • Suitable for fluorescent labeling of proteins, peptides, and other amine-containing biomolecules.
  • High supplied purity (98.0%).
  • Available in lab-scale and larger package sizes to support research workflows.