Learn More
Apexbio Technology LLC FITC Rabbit Anti-Goat IgG (H+L) Antibody 1ml
SDP
Catalog No. 5000096988
Encompass_Preferred
FITC Rabbit Anti-Goat IgG HplusL Antibody is a polyclonal secondary antibody targeting goat immunoglobulins recognizing both heavy and light chains Its mechanism of action involves specific binding to goat-derived primary antibodies enabling indirect detection and visualization of antigen-antibody complexes through conjugation with fluorescein isothiocyanate (FITC) The antibody is recommended for use in immunofluorescence microscopy (1 100 1 500) flow cytometry (1 200 1 1000) and fluorescence-based immunohistochemistry (1 50 1 200) Based on these characteristics FITC Rabbit Anti-Goat IgG HplusL Antibody is suitable for applications in fluorescence microscopy flow cytometry and immunohistochemistry research
Catalog No. 50-000-96988 Supplier Apexbio Technology LLC Supplier No. K12131
FITC Rabbit Anti-Goat IgG HplusL Antibody is a polyclonal secondary antibody targeting goat immunoglobulins recognizing both heavy and light chains Its mechanism of action involves specific binding to goat-derived primary antibodies enabling indirect detection and visualization of antigen-antibody complexes through conjugation with fluorescein isothiocyanate (FITC) The antibody is recommended for use in immunofluorescence microscopy (1 100 1 500) flow cytometry (1 200 1 1000) and fluorescence-based immunohistochemistry (1 50 1 200) Based on these characteristics FITC Rabbit Anti-Goat IgG HplusL Antibody is suitable for applications in fluorescence microscopy flow cytometry and immunohistochemistry research