Learn More
ABclonal Technology FITC Rabbit anti-Human CD99
SDP
Catalog No. 5000012532
Encompass_Preferred
The protein encoded by this gene is a cell surface glycoprotein involved in leukocyte migration T-cell adhesion ganglioside GM1 and transmembrane protein transport and T-cell death by a caspase-independent pathway In addition the encoded protein may have the ability to rearrange the actin cytoskeleton and may also act as an oncosuppressor in osteosarcoma This gene is found in the pseudoautosomal region of chromosomes X and Y and escapes X-chromosome inactivation There is a related pseudogene located immediately adjacent to this locus
Catalog No. 50-000-12532 Supplier ABclonal Technology Supplier No. A26936500T
The protein encoded by this gene is a cell surface glycoprotein involved in leukocyte migration T-cell adhesion ganglioside GM1 and transmembrane protein transport and T-cell death by a caspase-independent pathway In addition the encoded protein may have the ability to rearrange the actin cytoskeleton and may also act as an oncosuppressor in osteosarcoma This gene is found in the pseudoautosomal region of chromosomes X and Y and escapes X-chromosome inactivation There is a related pseudogene located immediately adjacent to this locus