Learn More
Biolegend FITC Rat IgG2a, κ Isotype Ctrl
Supplier:  Biolegend 400506
Encompass_Preferred
FITC Rat IgG2a, κ Isotype Ctrl [RTK2758]; Isotype: Rat IgG2a, κ; Reactivity: KLH; Apps: FC, ICFC; Size: 200 ug
Catalog No. NC9897274