Learn More
Immunology Consultants anti-Goat IgG h+l Antibody (Rabbit) - FITC Conjugated
SDP
Catalog No. 502248874
Encompass_Preferred
RGHL‑50F is a FITC‑conjugated rabbit antibody targeting goat IgG heavy and light chains, optimized for fluorescence‑based detection. The fluorescein label enables bright, clear visualization in microscopy, flow cytometry, and fluorescent immunoassays. Produced by Immunology Consultants and supplied in a 1 mg format, RGHL‑50F provides reliable, high‑specificity performance for fluorescence‑based secondary detection.
This item is not returnable. View return policy
Catalog No. 50-224-8874 Supplier Immunology Consultants Supplier No. RGHL50F
RGHL‑50F is a FITC‑conjugated rabbit antibody targeting goat IgG heavy and light chains, optimized for fluorescence‑based detection. The fluorescein label enables bright, clear visualization in microscopy, flow cytometry, and fluorescent immunoassays. Produced by Immunology Consultants and supplied in a 1 mg format, RGHL‑50F provides reliable, high‑specificity performance for fluorescence‑based secondary detection.