Learn More
Biolegend FITC anti-human CD4
Supplier:  Biolegend 357406
Encompass_Preferred
FITC anti-human CD4 [A161A1]; Isotype: Rat IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC0586095