Learn More
Diagnostic Biosystems FITE STAIN Fite's Stain
Catalog No. 502407550
Encompass_Preferred
The Fite’s Stain Kit (For Leprosy and Nocardia) is intended for use in the histological visualization of mycobacterium Lepra bacillus and Nocardia. This kit may be used on formalin-fixed, paraffin-embedded sections.\n
Catalog No. 50-240-7550 Supplier Diagnostic Biosystems Supplier No. KT013
The Fite’s Stain Kit (For Leprosy and Nocardia) is intended for use in the histological visualization of mycobacterium Lepra bacillus and Nocardia. This kit may be used on formalin-fixed, paraffin-embedded sections.\n