Learn More
Medchemexpress LLC HY-101845 5mg Medchemexpress, FITM CAS:932737-65-0 Purity:>98%
SDP
Catalog No. 501967886
Encompass_Preferred
Medchemexpress, HY-101845 5mg FITM CAS:932737-65-0 FITM is a negative allosteric modulator of mGlu1 receptor with a Ki of 2.5 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7886 Supplier Medchemexpress LLC Supplier No. HY1018455MG
Medchemexpress, HY-101845 5mg FITM CAS:932737-65-0 FITM is a negative allosteric modulator of mGlu1 receptor with a Ki of 2.5 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.