Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) PLCD20006 Quick-Disconnect Compression Insert; Valved, 3/8"" Tube OD, 1/4"" Flow Size
Catalog No. 50628096
Encompass
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.
Catalog No. 50-628-096 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0636075
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.