Learn More
Cole-Parmer FITTING 1/16OD TUBE,10-32CONED
Supplier:  Cole-Parmer 0201868
Encompass
Idex Fingertight One-Piece Fitting, ChromTRAC™, Multicolor PEEK, 1/16" OD Tubing, 10-32 Coned; 10/pk
Catalog No. NC2204832