Learn More
Waters Corp Fitting, 1/4-28, PEEK, Wash Port
Supplier: Waters Corp 700005297
This replacement fitting serves as a wash port fitting for various Waters ACQUITY systems. It features a a 1/4–28 thread and is constructed from PEEK, ensuring chemical compatibility and durability for demanding laboratory environments.
- Constructed from PEEK material
- Features a 1/4–28 thread
- Functions as a wash port fitting
- Compatible with ACQUITY UPLC, H-Class, H-Class Bio, I-Class, and ACQUITY Premier Sample Manager – FTN's
- Compatible with ACQUITY UPC2 SM-FL systems