Learn More
Waters Corp Waters Fitting, 1/4 Barbed, 1/8-27 NPT, Drip Tray Connection
Catalog No. 50179746 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This fitting is designed to connect a 1/4 barbed hose to a 1/8-27 NPT thread, commonly used for drip tray drain line connections.
  • Connects 1/4 barbed hose to 1/8-27 NPT thread.
  • Designed for drip tray drain line connections.
  • Easily installed into drip trays.
Catalog No. 50-179-746 Supplier Waters Corp Supplier No. 415000221
This fitting is designed to connect a 1/4 barbed hose to a 1/8-27 NPT thread, commonly used for drip tray drain line connections.
  • Connects 1/4 barbed hose to 1/8-27 NPT thread.
  • Designed for drip tray drain line connections.
  • Easily installed into drip trays.