Learn More
Waters Corp Waters Fitting, 6-40, One Piece, PEEK
Supplier:  Waters Corp 700002715
Encompass_Preferred
The one piece PEEK fitting is designed to fit into the 5000psi nanoACQUITY system.
  • One-piece PEEK fitting
  • Designed for the 5000psi nanoACQUITY system
  • Comes in a pack of 10
Catalog No. 50-792-790