Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, HDPE, Straight, Hose Barb to Threaded Adapter, 1/8"" ID x 1/4"" NPT(M); 10/PK
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3062142
Encompass
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb
Catalog No. 50-628-868