Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, HDPE, Straight, Hose Barb to Threaded Adapter, 3/8"" ID x 1/4"" NPT(M); 100/PK
Catalog No. 50628916
Encompass
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb
Catalog No. 50-628-916 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3062552
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb