Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Nylon, Straight, Hose Barb to Threaded Adapter, 1/4"" ID x 1/8"" NPT(M); 100/PK
Catalog No. 50628823
Encompass
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb.
Catalog No. 50-628-823 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3062532
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb.