Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Nylon, Straight, Hose Barb to Threaded Adapter, 5/32"" ID x 1/8"" NPT(M); 10/PK
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3062124
Encompass
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb.
Catalog No. 50-628-793