Learn More
AVANTOR MASTERFLEX (VWR LLC) FITTING ADAPT3/4FTFTX5/8FTFT 1
Catalog No. 502742306
Encompass
Masterflex Fitting, Polypropylene, Straight, Hosebarb to Thread, 5/8IN ID x 3/4IN NPT(M)10/PK, Avantor
Catalog No. 50-274-2306 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4062111
Masterflex Fitting, Polypropylene, Straight, Hosebarb to Thread, 5/8IN ID x 3/4IN NPT(M)10/PK, Avantor