Learn More
DSI LLC Fitting, black, PEEK-HT, one-piece, 10-32, 5/pkg, For UHPLC, up to 1000bar / 500bar as fingertight
Supplier:  DSI LLC JR550315
Encompass_Preferred
Fitting, black, PEEK-HT, one-piece, 10-32, 5/pkg, For UHPLC, up to 1000bar / 500bar as fingertight
Catalog No. 50-225-4807