Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) PLCD12006 Quick-Disconnect Panel-Mount Compression Body; Valve, 3/8""OD, Acetal; PLCD
Catalog No. 50628065
Encompass
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.
Catalog No. 50-628-065 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0636050
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.