Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) PLCD1200612 Quick-Disconnect Panel-Mount Compression Fitting Body; Valved, 3/8"" Tube OD, PP
Catalog No. 50628066
Encompass
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.
Catalog No. 50-628-066 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0636450
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.