Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) LCD10004 Metal quick-disconnect coupling body, valved, 1/4"" NPT (M) thread, 1/4"" flow size
Catalog No. 50627774
Encompass
Metal quick-disconnect couplings require a fitting body and an insert with the same flow size (order separately). Withstand vacuum to 250 psi, temperatures from -40 to 180¿F, and can be used with gases or liquids.
Catalog No. 50-627-774 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3140300
Metal quick-disconnect couplings require a fitting body and an insert with the same flow size (order separately). Withstand vacuum to 250 psi, temperatures from -40 to 180¿F, and can be used with gases or liquids.