Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) LCD17004 Coupling bodies; hose barb valved fittings; 1/4"" ID; 2.00"" length
Catalog No. 50627778
Encompass
Metal quick-disconnect couplings require a fitting body and an insert with the same flow size (order separately). Withstand vacuum to 250 psi, temperatures from -40 to 180¿F, and can be used with gases or liquids.
Catalog No. 50-627-778 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3140320
Metal quick-disconnect couplings require a fitting body and an insert with the same flow size (order separately). Withstand vacuum to 250 psi, temperatures from -40 to 180¿F, and can be used with gases or liquids.