Learn More
Perkin Elmer US LLC Female Barb CTFE Fitting
Catalog No. NC0883464
Encompass_Preferred
This fitting is included in the Sample Introduction Kit for NexION 2000 Productivity Configuration - Black (N8150036), Sample Introduction Kit for NexION 2000 Semi-Conductor Configuration -White (N8150037), NexION 300/350Q/X/D Consumables Kit (N8140513), NexION 300/350S Consumables Kit (N8140514), Single Cell Kit for NexION 350 (N8140032), Single Cell Kit for NexION 1000/2000 (N8150032), and the Nebulizer Liquid Connector Kit (N8145016).
Catalog No. NC0883464 Supplier Perkin Elmer US LLC Supplier No. N8145018
This fitting is included in the Sample Introduction Kit for NexION 2000 Productivity Configuration - Black (N8150036), Sample Introduction Kit for NexION 2000 Semi-Conductor Configuration -White (N8150037), NexION 300/350Q/X/D Consumables Kit (N8140513), NexION 300/350S Consumables Kit (N8140514), Single Cell Kit for NexION 350 (N8140032), Single Cell Kit for NexION 1000/2000 (N8150032), and the Nebulizer Liquid Connector Kit (N8145016).