Learn More
AVANTOR MASTERFLEX (VWR LLC) Cadence 316 SS fittings; female luer x 10-32 UNF thread 41507-33
Catalog No. NC0538354
Encompass
Exceptional durability. Easy to disassemble for cleaning. Ideal for harsh chemical environments. Autoclavable.
Catalog No. NC0538354 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4150733
Exceptional durability. Easy to disassemble for cleaning. Ideal for harsh chemical environments. Autoclavable.