Learn More
Grainger FOAMGLAS Pipe Fitting Insulation, 90° Elbow, Black, 1 in. Thick, 1 ea
Catalog No. 502782467
Encompass_Preferred
FOAMGLAS® cellular glass insulation is a lightweight, rigid, and durable material engineered from millions of completely sealed glass cells. Designed for industrial insulation, this inorganic, closed-cell glass product provides reliable thermal efficiency for systems including cryogenic, chilled water, and hydrocarbon processing applications. It remains dimensionally stable over time, resisting common industrial chemicals, moisture, and corrosion, while offering a non-combustible and vermin-proof barrier.
  • Incombustible material with 0 flame spread index
  • Exceptional water and vapor tightness for long-lasting performance
  • Resistant to most common industrial chemicals and acids
  • Dimensionally stable with no shrinkage or swelling
  • Contains no flame retardants or blowing agents with high global warming potential
  • Service temperature range of -450°F to +900°F
Catalog No. 50-278-2467 Supplier Grainger Supplier No. 19NG67
FOAMGLAS® cellular glass insulation is a lightweight, rigid, and durable material engineered from millions of completely sealed glass cells. Designed for industrial insulation, this inorganic, closed-cell glass product provides reliable thermal efficiency for systems including cryogenic, chilled water, and hydrocarbon processing applications. It remains dimensionally stable over time, resisting common industrial chemicals, moisture, and corrosion, while offering a non-combustible and vermin-proof barrier.
  • Incombustible material with 0 flame spread index
  • Exceptional water and vapor tightness for long-lasting performance
  • Resistant to most common industrial chemicals and acids
  • Dimensionally stable with no shrinkage or swelling
  • Contains no flame retardants or blowing agents with high global warming potential
  • Service temperature range of -450°F to +900°F