This cellular glass 90° elbow is designed for high-performance thermal insulation in demanding industrial piping systems. Constructed from rigid, closed-cell glass, it provides consistent thermal resistance and maintains structural integrity in environments ranging from cryogenic temperatures to high-heat applications. The fitting features an anti-abrasive coating and factory-applied external protection, ensuring durability and ease of installation.
- Provides an R-value of 5.2 for efficient thermal management
- Operates across a wide temperature range of -450°F to +900°F
- Resists moisture, mold, mildew, and corrosion for long-term reliability
- Suitable for environments including chemical processing, chilled water, and hydrocarbon systems
- Plenum rated and compliant with ASTM C 1639 standards
- Features rigid construction with a closed-cell structure