Learn More
Grainger FOAMGLAS Pipe Fitting Insulation, 90° Elbow, Cellular Glass, 2 in. ID, 2 in. Thick
Catalog No. 502778741
Encompass_Preferred
This cellular glass insulation is designed for 90° elbow pipe fittings, providing a rigid and efficient thermal solution. Factory trimmed and preformed, these half-sections are engineered to simplify installation and reduce field fabrication time. It is highly resistant to extreme temperatures, moisture, and corrosive environments, making it suitable for a wide range of industrial piping systems.
  • Operating temperature range from -450°F to +900°F
  • 6.9 R-Value for effective thermal performance
  • Resistant to corrosion, moisture, mold, and microbes
  • Closed-cell structure prevents fluid and pest infiltration
  • Plenum rated for safety
  • Fits 1 1/2 in. nominal pipe size with 2 in. wall thickness
  • Complies with ASTM E84, E136, and E96 standards
Catalog No. 50-277-8741 Supplier Grainger Supplier No. 19NG89
This cellular glass insulation is designed for 90° elbow pipe fittings, providing a rigid and efficient thermal solution. Factory trimmed and preformed, these half-sections are engineered to simplify installation and reduce field fabrication time. It is highly resistant to extreme temperatures, moisture, and corrosive environments, making it suitable for a wide range of industrial piping systems.
  • Operating temperature range from -450°F to +900°F
  • 6.9 R-Value for effective thermal performance
  • Resistant to corrosion, moisture, mold, and microbes
  • Closed-cell structure prevents fluid and pest infiltration
  • Plenum rated for safety
  • Fits 1 1/2 in. nominal pipe size with 2 in. wall thickness
  • Complies with ASTM E84, E136, and E96 standards